  • Search Results
  • Does bromocriptine play a role in decreasing oxidative stress for early weaned programmed obesity?
Does bromocriptine play a role in decreasing oxidative stress for early weaned programmed obesity?
Life sciences (2013-12-24)
Nayara Peixoto-Silva, Ellen P S Conceição, Janaine C Carvalho, Natália S Lima, José Firmino Nogueira-Neto, Elaine de Oliveira, Egberto G Moura, Patricia C Lisboa
ABSTRACT
Studies have demonstrated that early weaning can promote metabolic syndrome during adulthood and that obesity increases oxidative stress. Thus, we aimed to evaluate redox status in a pharmacological early weaning rodent model programmed for metabolic syndrome at adulthood. Lactating dams were randomly assigned into 2 groups: the early weaning group (BRO), which was treated intraperitoneally with bromocriptine (1 mg/day) to inhibit prolactin secretion for the last 3 days of lactation, and the control group (C), which received the BRO diluent for the same time period. The offspring were killed at 90 (PN90) and 180 (PN180) days after birth. Early weaning induced greater visceral adiposity and dyslipidemia. At PN90, the BRO offspring showed glucose intolerance with normoinsulinemia and increased plasma and liver superoxide dismutase, and liver glutathione peroxidase activities, which reduced the liver malondialdehyde but not the increased plasma malondialdehyde levels. However, the BRO offspring showed insulin resistance at PN180 and increased plasma glutathione peroxidase, liver superoxide dismutase, and catalase activities. These changes reduced the plasma and liver malondialdehyde levels, which aided in hepatocyte architecture preservation. Additionally, we observed that sirtuin 1 was overexpressed in the BRO group at PN90, but the increased expression was not maintained through PN180, which suggests unfavorable metabolic conditions in the older offspring. Despite the observed obesity and glucose homeostasis dysfunction, our data suggest that the early weaning programming induced by bromocriptine can improve the offspring's redox status and may prevent liver damage during adulthood.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Prolactin human, recombinant, expressed in E. coli, lyophilized powder, BioReagent, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
2-Bromo-α-ergocryptine methanesulfonate salt, solid
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Bromocriptine mesilate for system suitability, European Pharmacopoeia (EP) Reference Standard