MilliporeSigma
  • Prognostic significance of HIF-1a, CA-IX, and OPN in T1-T2 laryngeal carcinoma treated with radiotherapy.
Prognostic significance of HIF-1a, CA-IX, and OPN in T1-T2 laryngeal carcinoma treated with radiotherapy.
The Laryngoscope (2013-10-30)
Jan E Wachters, Michiel L Schrijvers, Lorian Slagter-Menkema, Mirjam Mastik, Geertruida H de Bock, Johannes A Langendijk, Philip M Kluin, Ed Schuuring, Bernard F A M van der Laan, Jacqueline E van der Wal
ABSTRACT
To examine the prognostic value of hypoxia inducible factor HIF-1a, CA-IX, and OPN on clinical outcome in patients with T1-T2 supraglottic laryngeal squamous cell carcinoma (LSCC) treated with primarily radiotherapy (RT). Retrospective cohort study. Tumor tissue sections of 60 patients with T1-T2 supraglottic LSCC treated with primarily radiotherapy were assessed immunohistochemically for expression of HIF-1a, CA-IX, and OPN. The relationship of protein expression and classical clinical parameters with clinical outcome was studied, using Cox regression and Kaplan-Meier survival analyses. Neither HIF-1a nor CA-IX was of prognostic significance toward local control or overall survival in T1-T2 supraglottic LSCC. Cox regression survival analysis showed no relation between HIF-1a or CA-IX expression and local control (HR [hazard ratio] 1.07, CI [95% confidence interval] 0.29-3.87; HR 0.34, CI 0.04-2.58). Furthermore, OPN expression was not associated with local control (HR 1.37, CI 0.45-4.17) and overall survival (HR 0.99, CI 0.44-2.21). Our earlier findings in T1-T2 glottic LSCC (Schrijvers et al., 2008) could not be confirmed. The absence of prognostic significance for HIF-1a and CA-IX toward local control in supraglottic LSCC, unlike glottic LSCC, suggests that supraglottic LSCC might represent another biological entity.
Sigma-Aldrich
Carbonic Anhydrase II human, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase II bovine, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, ≥5,000 units/mg protein, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6