  • Search Results
  • Role of carbonic anhydrases in pathogenic micro-organisms: a focus on Aspergillus fumigatus.
Role of carbonic anhydrases in pathogenic micro-organisms: a focus on Aspergillus fumigatus.
Journal of medical microbiology (2013-10-24)
Jaqueline Moisés Tobal, Márcia Eliana da Silva Ferreira Balieiro
ABSTRACT
Aspergillus fumigatus is a ubiquitous saprophytic fungus responsible for organic material decomposition, and plays an important role in recycling environmental carbon and nitrogen. Besides its important role in the environment, this fungus has been reported as one of the most important fungal pathogens in immunocompromised patients. Due to changes in CO2 concentration that some pathogens face during the infection process, studies have been undertaken to understand the pathogenic roles of carbonic anhydrases (CAs), well-known CO2 hydration catalytic enzymes. As a basis for a discussion of the possible roles of CAs in A. fumigatus pathogenicity, this review describes the main characteristics of the A. fumigatus infection and the challenges for its treatment. In addition, it gathers findings from studies with CA inhibitor drugs as anti-infective agents in different pathogens.
Product Description
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase II human, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase II bovine, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, ≥5,000 units/mg protein, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4