  • Search Results
  • Establishment of cell suspension cultures of two Costa Rican Jatropha species (Euphorbiaceae).
Establishment of cell suspension cultures of two Costa Rican Jatropha species (Euphorbiaceae).
Revista de biologia tropical (2013-09-14)
Laura Yesenia Solís-Ramos, Laura Miranda Carballo, Marta Valdez-Melara
ABSTRACT
J. curcas has been studied in different countries and some interesting agronomic, pharmacological and industrial properties have been reported. More recently, it has been considered an important alternative source for biofuel production. The objective of this study was to establish a long-term method for the maintenance of calli and cell suspension cultures of the local species J. curcas and J. gossypifolia, in order to allow future studies for novel compounds with pharmaceutical or industrial applications. For this, friable calli were successfully induced from hypocotyl segments of.. curcas and J. gossypifolia that were cultured in semisolid MS media supplemented with 1.5 mg/L, and 0.5 mg/L of 2,4-D, respectively. Cell suspension cultures of J. curcas were established using 1 g of 35 and 60-day calli, in 50 mL of liquid MS media supplied with 1.5 mg/L of 2,4-D; sucrose and maltose were additionally evaluated as carbon sources. After 35 days, cell suspension cultures initiated with 35-day calli, showed greater cell growth with a maximum biomass of 194.9 g/L fresh weight, 6.59 g/L dry weight and 17.3% packed volume. The exponential phase ended at day 35 for cultures initiated with 35-day calli, and at day 21 for cultures initiated with 60-day calli. Higher biomass production was obtained with sucrose. Cell cultures were established with 35-day calli in MS media with the same 2,4-D concentration used for calli induction and 30g/L sucrose. This medium was considered optimum for the maintenance and growth of cell suspensions for both species, with sub-cultures every 20 days. The biotechnological potential for the production of bioactive compounds in these species for pharmacological, agricultural and industrial applications is being evaluated.
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D solution, certified reference material, 5000 μg/mL in acetonitrile
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D, certified reference material, TraceCERT®