  • Search Results
  • Significant variation of immunohistochemical marker expression in paired primary and metastatic clear cell renal cell carcinomas.
Significant variation of immunohistochemical marker expression in paired primary and metastatic clear cell renal cell carcinomas.
American journal of clinical pathology (2013-08-21)
Zenggang Pan, William Grizzle, Omar Hameed
ABSTRACT
To compare the immunohistochemical expression of diagnostic markers in primary clear cell renal cell carcinomas (RCCs) and their matched metastases. Tissue microarrays were constructed from 15 pairs of primary and metastatic clear cell RCCs and then evaluated for the immunohistochemical expression of renal cell carcinoma antigen (RCCA), kidney-specific cadherin, carbonic anhydrase IX (CAIX), and paired box genes 2 (PAX2) and 8 (PAX8). There was significantly higher overall marker expression in metastatic tumors compared to their matched primaries (P < .001). Individually, there was greater CAIX, PAX2, and PAX8 expression and lower RCCA expression in metastatic tumors. Most importantly, a significant proportion of originally RCCA-positive tumors lost such expression in metastases. Metastatic RCCs have significantly higher expression of PAX2 and PAX8 compared to primary RCCs. RCCA is not very reliable in this diagnostic setting, both because of its lower overall sensitivity and loss of expression in metastatic RCCs.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder