  • Search Results
  • Electroconvulsive therapy for psychosis in a patient with epilepsy related to hypothalamic hamartoma.
Electroconvulsive therapy for psychosis in a patient with epilepsy related to hypothalamic hamartoma.
Epileptic disorders : international epilepsy journal with videotape (2013-08-01)
Elisabeth Ruppert, Laurence Lalanne, Jack Foucher, Marie-Agathe Zimmermann, Edouard Hirsch, Pierre Vidailhet
ABSTRACT
Psychosis is more common in people with temporal lobe epilepsy than it is in the general population. Treatment can be difficult in these patients because of the complex interactions between antipsychotic and antiepileptic drugs. Some antipsychotic drugs also decrease the seizure threshold. We report the case of a 49-year-old man with a hypothalamic hamartoma, with a history of both gelastic and temporal lobe seizures. The patient was rendered seizure-free after three neurosurgical procedures but developed a drug-resistant paranoid psychosis. He was treated with electroconvulsive therapy (ECT). After two weeks with six stimulations that resulted in seizures, the psychiatric phenomena disappeared completely. There was no relapse of either the psychiatric symptoms or the seizures during the 42 months of follow-up. This case report suggests that ECT might be safe for psychosis in patients with a history of seizures that have previously been successfully treated with neurosurgery, although caution should be exercised in drawing general conclusions from a single case report.
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Lamotrigine for system suitability, European Pharmacopoeia (EP) Reference Standard
Lamotrigine for peak identification, European Pharmacopoeia (EP) Reference Standard
Lamotrigine, European Pharmacopoeia (EP) Reference Standard
Supelco
Lamotrigine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®