  • Search Results
  • Asbestos-induced alveolar epithelial cell apoptosis. The role of endoplasmic reticulum stress response.
Asbestos-induced alveolar epithelial cell apoptosis. The role of endoplasmic reticulum stress response.
American journal of respiratory cell and molecular biology (2013-07-28)
David W Kamp, Gang Liu, Paul Cheresh, Seok-Jo Kim, Amanda Mueller, Anna P Lam, Humberto Trejo, David Williams, Sandhya Tulasiram, Margaret Baker, Karen Ridge, Navdeep S Chandel, Rohinee Beri
ABSTRACT
Asbestos exposure results in pulmonary fibrosis (asbestosis) and malignancies (bronchogenic lung cancer and mesothelioma) by mechanisms that are not fully understood. Alveolar epithelial cell (AEC) apoptosis is important in the development of pulmonary fibrosis after exposure to an array of toxins, including asbestos. An endoplasmic reticulum (ER) stress response and mitochondria-regulated (intrinsic) apoptosis occur in AECs of patients with idiopathic pulmonary fibrosis, a disease with similarities to asbestosis. Asbestos induces AEC intrinsic apoptosis, but the role of the ER is unclear. The objective of this study was to determine whether asbestos causes an AEC ER stress response that promotes apoptosis. Using human A549 and rat primary isolated alveolar type II cells, amosite asbestos fibers increased AEC mRNA and protein expression of ER stress proteins involved in the unfolded protein response, such as inositol-requiring kinase (IRE) 1 and X-box-binding protein-1, as well as ER Ca²(2+) release ,as assessed by a FURA-2 assay. Eukarion-134, a superoxide dismutase/catalase mimetic, as well as overexpression of Bcl-XL in A549 cells each attenuate asbestos-induced AEC ER stress (IRE-1 and X-box-binding protein-1 protein expression; ER Ca²(2+) release) and apoptosis. Thapsigargin, a known ER stress inducer, augments AEC apoptosis, and eukarion-134 or Bcl-XL overexpression are protective. Finally, 4-phenylbutyric acid, a chemical chaperone that attenuates ER stress, blocks asbestos- and thapsigargin-induced AEC IRE-1 protein expression, but does not reduce ER Ca²(2+) release or apoptosis. These results show that asbestos triggers an AEC ER stress response and subsequent intrinsic apoptosis that is mediated in part by ER Ca²(2+) release.
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Sigma-Aldrich
Sodium phenylbutyrate, ≥98% (HPLC)