MilliporeSigma
  • Novel taste-masked orally disintegrating tablets for a highly soluble drug with an extremely bitter taste: design rationale and evaluation.
Novel taste-masked orally disintegrating tablets for a highly soluble drug with an extremely bitter taste: design rationale and evaluation.
Drug development and industrial pharmacy (2012-09-22)
Qunyou Tan, Li Zhang, Guodong Liu, Dan He, Huafeng Yin, Hong Wang, Jianyong Wu, Hong Liao, Jingqing Zhang
ABSTRACT
The purpose of this study was to evaluate the taste masking potential of novel solid dispersions (SDs) using Eudragit® EPO as the excipient when incorporated into the orally disintegrating tablets (ODTs) for delivering a highly soluble drug with an extremely bitter taste. The pyridostigmine bromides (PB) SDs (PBSDs) were prepared by solvent evaporation-deposition method. The physicochemical properties of PBSDs were investigated by means of differential scanning calorimetry and Fourier transformed infrared spectroscopy. The dissolution test showed that only about 8% of PB was released from PBSDs in the simulated salivary fluid in 30 s. Therefore, PBSDs were considered taste-masked and selected for formulation of PBODTs. A central composite design was employed for process optimization. Multiple linear regression analysis for process optimization revealed that the optimal PBODTs were obtained, when the microcrystalline cellulose and crospovidone were 17.16 and 5.55 (%, w/w), respectively, and the average in vivo disintegration time was 25 s. The bitterness threshold of PB was examined by a sensory test, and the threshold value was set as 3 mg in each tablet. Taste evaluation of PBODTs in 18 volunteers revealed considerable taste masking with bitterness below the threshold value. PBODTs also revealed rapid drug release (around 99%, 2 min) in the simulated gastric fluid. The mean PB plasma concentration-time profiles of PBODTs and that of the commercial tablets were comparable, with closely similar pattern. Bioequivalence assessment results demonstrated that PBODTs and the commercial tablets were bioequivalent. In conclusion, PBODTs are prepared successfully, with taste masking and rapid disintegration in the oral cavity.
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Pyridostigmine bromide
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Kollidon® 25
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Povidone, European Pharmacopoeia (EP) Reference Standard
Pyridostigmine bromide, European Pharmacopoeia (EP) Reference Standard
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size