  • Search Results
  • Relative quantitative expression of hypoxia-inducible factor 1α messenger ribonucleic acid in recurrent craniopharyngiomas.
Relative quantitative expression of hypoxia-inducible factor 1α messenger ribonucleic acid in recurrent craniopharyngiomas.
Neurology India (2014-03-13)
Hao Liu, Zhiyong Liu, Jin Li, Qiang Li, Chao You, Jianguo Xu
ABSTRACT
Craniopharyngioma is a benign tumor, but recurrences are common and prognosis is poor. The pathologic mechanism underlying the high recurrence is still unknown. We hypothesized that there are hypoxic microenvironments within craniopharyngiomas and hypoxia inducible factor-1 alpha (HIF-1α) and its related genes are largely expressed in recurrent craniopharyngiomas. A total of 19 patients with craniopharyngiomas have been identified. The relative quantitative expressions of HIF-1α, vascular endothelial growth factor (VEGF) and carbonic adhydrase 9 (CA9) messenger ribonucleic acid (mRNA) of craniopharyngioma tissues were detected by real time reverse transcription polymerase chain reaction. HIF-1α and VEGF mRNA was significantly up-regulated in recurrent craniopharyngiomas. Mean expression levels (recurrent craniopharyngiomas compared with non-recurrent craniopharyngiomas, as normalized to expression of β-actin) were 3.09 versus 0.75 (P = 0.001) for HIF-1α, 1.07 versus 0.32 (P = 0.000) for VEGF, 1.21 versus 1.93 (P = 0.503) for CA9. In craniopharyngiomas, the expression of VEGF showed a significant correlation with HIF-1α (r = 0.836, P = 0.000). There were hypoxic microenvironments within craniopharyngiomas. Therefore, preventing the tumor cells from adapting to the hypoxic conditions may be an effective way to obviate the relapse of craniopharyngioma.
Product Description
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 10 purifications
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase II bovine, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, ≥5,000 units/mg protein, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase II human, recombinant, expressed in E. coli, buffered aqueous solution