  • Search Results
  • Quantitative estimation of sialic acids. II. A colorimetric resorcinol-hydrochloric acid method.
Quantitative estimation of sialic acids. II. A colorimetric resorcinol-hydrochloric acid method.
Biochimica et biophysica acta (1957-06-01)
L SVENNERHOLM
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis