  • Search Results
  • Identification of signal bias in the variable flip angle method by linear display of the algebraic Ernst equation.
Identification of signal bias in the variable flip angle method by linear display of the algebraic Ernst equation.
Magnetic resonance in medicine (2011-03-25)
Gunther Helms, Henning Dathe, Nikolaus Weiskopf, Peter Dechent
ABSTRACT
A novel linear parameterization for the variable flip angle method for longitudinal relaxation time T(1) quantification from spoiled steady state MRI is derived from the half angle tangent transform, τ, of the flip angle. Plotting the signal S at coordinates x=Sτ and y=S/τ, respectively, establishes a line that renders signal amplitude and relaxation term separately as y-intercept and slope. This representation allows for estimation of the respective parameter from the experimental data. A comprehensive analysis of noise propagation is performed. Numerical results for efficient optimization of longitudinal relaxation time and proton density mapping experiments are derived. Appropriate scaling allows for a linear presentation of data that are acquired at different short pulse repetition times, TR < T1 thus increasing flexibility in the data acquisition by removing the limitation of a single pulse repetition time. Signal bias, like due to slice-selective excitation or imperfect spoiling, can be readily identified by systematic deviations from the linear plot. The method is illustrated and validated by 3T experiments on phantoms and human brain.
Product Description
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, Type A, ash 5-6%
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type E, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Millipore
Agar, for chromogenic media
Millipore
Agar, high purity, low ionic content, low gel strenght
Millipore
Agar, for membrane filtration, low gel strengh