  • Search Results
  • COPD results in a reduction in UCP3 long mRNA and UCP3 protein content in types I and IIa skeletal muscle fibers.
COPD results in a reduction in UCP3 long mRNA and UCP3 protein content in types I and IIa skeletal muscle fibers.
Journal of cardiopulmonary rehabilitation (2004-12-17)
Aaron P Russell, Emmanuel Somm, Richard Debigaré, Oliver Hartley, Denis Richard, Giacomo Gastaldi, Astrid Melotti, Annie Michaud, Jean-Paul Giacobino, Patrick Muzzin, Pierre LeBlanc, François Maltais
ABSTRACT
Findings recently have shown coupling protein-3 (UCP3) content to be decreased in the skeletal muscle of patients with chronic obstructive pulmonary disease (COPD). Uncoupling protein-3 mRNA exists as two isoforms: long (UCP3L) and short (UCP3S). The UCP3 protein is expressed the least in oxidative and the most in glycolytic muscle fibers. Levels of UCP3 have been associated positively with intramyocellular triglyceride (IMTG) contents in conditions of altered fatty acid metabolism. As a source for muscle free fatty acid metabolism, IMTG is decreased in COPD. The current study completely characterized all the parameters of UCP3 expression (ie, UCP3L and UCP3S mRNA expression in whole muscle samples) and UCP3 protein content as well as IMTG content in the different fiber types in patients with COPD and healthy control subjects. Using real-time polymerase chain reaction, UCP3 gene expression was quantified. Skeletal muscle fiber type and UCP3 protein and IMTG content were measured using immunofluorescence and Oil red oil staining, respectively. The findings showed that UCP3L mRNA expression was 44% lower (P < .005) in the patients with COPD than in the control subjects, whereas the UCP3S mRNA content was similar in the two groups. As compared with control subjects, UCP3 protein content was decreased by 89% and 83% and the IMTG content by 64% and 54%, respectively, in types I and IIa fibers (P < .0167) of patients with COPD, whereas they were unchanged in IIx fibers. The reduced UCP3 and IMTG content in the more oxidative fibers may be linked to the altered muscle fatty acid metabolism associated with COPD. Further studies are required to determine the exact role and clinical relevance of the reduced UCP3 content in patients with COPD.
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, analytical standard
Supelco
Oleic acid, Selectophore, ≥99.0%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent