  • Search Results
  • Combination therapy of dyslipidemia in non-insulin-dependent diabetes mellitus and the metabolic syndrome.
Combination therapy of dyslipidemia in non-insulin-dependent diabetes mellitus and the metabolic syndrome.
Current diabetes reports (2004-10-06)
Christopher M Rembold
ABSTRACT
Non-insulin-dependent diabetes mellitus (NIDDM) and the metabolic syndrome separately and additively increase the risk for atherosclerotic cardiovascular disease. Considering the high cardiovascular risk associated with NIDDM and the metabolic syndrome, aggressive therapy of dyslipidemia with tailored combination therapy should be considered given informed consent and discussion of risks. In addition to statins, niacin, and fibrates, therapies shown to decrease the risk for atherosclerotic cardiovascular disease include omega-3 fatty acids, diet, exercise, and optimal blood pressure control with thiazides and blockers of the renin-angiotensin system. These therapies should also be considered to reduce the high cardiovascular risk associated with NIDDM and the metabolic syndrome.
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Supelco
Nicotinamide (Niacinamide), analytical standard