Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, DARCO® KB-G
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder