Dietary intake of vitamin D is not enough.
The practising midwife (2013-08-06)
Valerie Hemmings
ABSTRACT
The National Diet and Nutrition Survey (Bates et al 2011) show adult females have low levels of vitamin D. Many women in midwives' care may be vitamin D deficient. The aim of this article is to update midwives' knowledge of vitamin D in line with their statutory role as a midwife, so they can recognise deficiency and make women aware of the consequences for themselves and their baby.
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholecalciferol (D3), analytical standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, analytical standard