  • Search Results
  • Cellular uptake and activity of heparin functionalised cerium oxide nanoparticles in monocytes.
Cellular uptake and activity of heparin functionalised cerium oxide nanoparticles in monocytes.
Biomaterials (2013-03-13)
S R Simon Ting, John M Whitelock, Romana Tomic, Cindy Gunawan, Wey Yang Teoh, Rose Amal, Megan S Lord
ABSTRACT
Cerium oxide nanoparticles (nanoceria) are effective in scavenging intracellular reactive oxygen species (ROS). In this study nanoceria synthesized by flame spray pyrolysis (dXRD = 12 nm) were functionalised with heparin via an organosilane linker, 3-aminopropyltriethoxysilane. Nanoceria were functionalised with approximately 130 heparin molecules per nanoparticle as determined by thermo gravimetric analysis. Heparin functionalised nanoceria were more effectively internalised by the human monocyte cell line, U937, and U937 cells that had been activated with phorbol 12 myristate 13-acetate (PMA) than bare nanoceria. The heparin functionalised nanoceria were also more effective in scavenging ROS than nanoceria in both activated and unactivated U937 cells. Heparin coupled nanoceria were found to be biologically active due to their ability to bind fibroblast growth factor 2 and signal through FGF receptor 1. Additionally, the heparin-coupled nanoceria, once internalised by the cells, were found to be degraded by 48 h. Together these data demonstrated that heparin enhanced the biological properties of nanoceria in terms of cellular uptake and ROS scavenging, while the nanoceria themselves were more effective at delivering heparin intracellularly than exposing cells to heparin in solution.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
Cerium(IV) oxide, dispersion, nanoparticles, <25 nm particle size, 10 wt. % in H2O
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, dispersion, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
Sigma-Aldrich
Cerium(IV) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Cerium, powder, −40 mesh, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, ≥99.0%
Sigma-Aldrich
Cerium, chips