  • Search Results
  • The effect of apricots on the experimental cataract model formed by sodium selenite.
The effect of apricots on the experimental cataract model formed by sodium selenite.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2013-01-26)
Selim Doganay, Cem Duz, Penpe Gul Firat, Cem Cankaya, Derya Kutukde, Yılmaz Cigremis
ABSTRACT
This study was designed in order to investigate whether sun dried apricots have a preventive effect on the experimental cataract model formed by sodium selenite in rats. Fifty-nine Spraque-Dawley rat pups were divided into three groups. Group 1 (control group) consisted of twenty rat pups, born from the rats nourished ad libitum. Group 2 consisted of 18 newborn rats, born from the rats nourished ad libitum with 10% sun dried natural apricots. Group 3 consisted of 21 newborn rats, born from the rats nourished ad libitum. Subcutaneous (30nmol/gr) sodium selenite injection was applied to all the newborn rats except the control group (Group 1) on postpartum day 10. Cataract development was graded by slit-lamp examination and photography. Encapsulated lenses were analyzed for reduced glutathione (GSH) and malondialdehyde (MDA), a marker of lipid per oxidation. Lenses were also analyzed for total nitrite (TN). The presence of oxidative stress in selenite cataract development and its prevention by sun dried apricots.
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%