MilliporeSigma
  • Single-dose pharmacokinetics and tolerability of oral delta-9- tetrahydrocannabinol in patients with amyotrophic lateral sclerosis.
Single-dose pharmacokinetics and tolerability of oral delta-9- tetrahydrocannabinol in patients with amyotrophic lateral sclerosis.
Drug metabolism letters (2012-05-19)
Markus Joerger, Justin Wilkins, Stefania Fagagnini, Reto Baldinger, Rudolf Brenneisen, Ursula Schneider, Bea Goldman, Markus Weber
ABSTRACT
Cannabinoids exert neuroprotective and symptomatic effects in amyotrophic lateral sclerosis (ALS). We assessed the pharmacokinetics (PK) and tolerability of delta-9-tetrahydrocannabinol (THC) in ALS patients. Nine patients received THC single oral doses of 5mg and 10mg, separated by a wash-out period of two weeks. Blood samples for the determination of THC, 11-nor-9-carboxy-THC (THC-COOH) and hydroxy-THC (THC-OH) were taken up to 8 hours after intake. Adverse events were assessed by visual analogue scales (VAS). Plasma concentrations of the active metabolite THC-OH were submitted to sequential pharmacokinetic-pharmacodynamic population modeling on individual heart rate as a proxy for THC's cardiovasculatory effects. Drowsiness, euphoria, orthostasis, sleepiness, vertigo and weakness were significantly more frequent in patients receiving 10mg compared to 5 mg THC. A marked interindividual variability was found for the absorption of oral THC (84%) and elimination of THC-COOH (45%). PK data did not support any clinically relevant deviation from linear PK in the investigated range of concentrations. Plasma concentrations of THC-OH were positively correlated with the individual heart rate. An E(max-model) was successfully fitted to individual heart rate, with a THC-OH plasma concentration of 3.2 x 10(-4) μmol/L for EC(50) and an E(max) of 93 bpm for heart rate. The higher 10mg dose of THC was dose-limiting in patients with ALS. High interindividual PK variability requires individuell titration of THC for potential therapeutic use in patients with ALS.
Product Description
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-11-Hydroxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-11-Hydroxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®