  • Search Results
  • Simultaneous quantification of Delta9-tetrahydrocannabinol, 11-hydroxy-Delta9-tetrahydrocannabinol, and 11-nor-Delta9-tetrahydrocannabinol-9-carboxylic acid in human plasma using two-dimensional gas chromatography, cryofocusing, and electron impact-mass spectrometry.
Simultaneous quantification of Delta9-tetrahydrocannabinol, 11-hydroxy-Delta9-tetrahydrocannabinol, and 11-nor-Delta9-tetrahydrocannabinol-9-carboxylic acid in human plasma using two-dimensional gas chromatography, cryofocusing, and electron impact-mass spectrometry.
Journal of chromatography. A (2007-07-21)
Ross H Lowe, Erin L Karschner, Eugene W Schwilke, Allan J Barnes, Marilyn A Huestis
ABSTRACT
A two-dimensional (2D) gas chromatography/electron impact-mass spectrometry (GC/EI-MS) method for simultaneous quantification of Delta(9)-tetrahydrocannabinol (THC), 11-hydroxy-Delta(9)-tetrahydrocannabinol (11-OH-THC), and 11-nor-Delta(9)-tetrahydrocannabinol-9-carboxylic acid (THCCOOH) in human plasma was developed and validated. The method employs 2D capillary GC and cryofocusing for enhanced resolution and sensitivity. THC, 11-OH-THC, and THCCOOH were extracted by precipitation with acetonitrile followed by solid-phase extraction. GC separation of trimethylsilyl derivatives of analytes was accomplished with two capillary columns in series coupled via a pneumatic Deans switch system. Detection and quantification were accomplished with a bench-top single quadrupole mass spectrometer operated in electron impact-selected ion monitoring mode. Limits of quantification (LOQ) were 0.125, 0.25 and 0.125 ng/mL for THC, 11-OH-THC, and THCCOOH, respectively. Accuracy ranged from 86.0 to 113.0% for all analytes. Intra- and inter-assay precision, as percent relative standard deviation, was less than 14.1% for THC, 11-OH-THC, and THCCOOH. The method was successfully applied to quantification of THC and its 11-OH-THC and THCCOOH metabolites in plasma specimens following controlled administration of THC.
Product Description
Supelco
(±)-11-Hydroxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-11-Hydroxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®