  • Search Results
  • Novel conformationally restricted tetracyclic analogs of delta8-tetrahydrocannabinol.
Novel conformationally restricted tetracyclic analogs of delta8-tetrahydrocannabinol.
Bioorganic & medicinal chemistry letters (1999-08-28)
A D Khanolkar, D Lu, P Fan, X Tian, A Makriyannis
ABSTRACT
Novel analogs of (-)-delta8-tetrahydrocannabinol (delta8-THC) in which the conformation of the side chain was restricted by incorporating the first one or two carbons into a six membered ring fused with the aromatic phenolic A ring were synthesized. The affinities of the novel ligands for CB1 and CB2 indicated that the "southbound" chain conformer retained the highest affinity for both receptors.
Product Description
Supelco
(−)-Δ8-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®