  • Search Results
  • [Action of the salts of univalent thallium on the liver mitochondria of the rat].
[Action of the salts of univalent thallium on the liver mitochondria of the rat].
Zhurnal evoliutsionnoi biokhimii i fiziologii (1984-07-01)
I A Skul'skiĭ, T I Ivanova, M V Savina
ABSTRACT
Using light scattering and electron microscopic techniques, studies have been made of the swelling of liver mitochondria in isotonic sucrose solutions containing different concentrations of nitrate or acetate of monovalent thallium. It was concluded that the injurious effect of thallium on the mitochondria is associated with both its penetration into intramitochondrial space, and interaction of the external thallium with mitochondrial membranes.
Sigma-Aldrich
Thallium(I) acetate, ≥99%
Sigma-Aldrich
Thallium(I) nitrate, 99.9% trace metals basis
Sigma-Aldrich
Thallium(I) nitrate, 99.999% trace metals basis
Sigma-Aldrich
Thallium(I) acetate, purum, ≥97.0%
Sigma-Aldrich
Thallium(III) acetate
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.