  • Search Results
  • Species relationships for plasma angiotensin converting enzyme activity using a furanacryloyl tripeptide substrate.
Species relationships for plasma angiotensin converting enzyme activity using a furanacryloyl tripeptide substrate.
Jikken dobutsu. Experimental animals (1989-04-01)
G O Evans
ABSTRACT
Angiotensin converting enzyme (ACE; EC 3.4.15.1) activities were compared in plasma samples obtained from three species using a furanacryloyl tripeptide substrate. The enzyme activity observed in Wistar rat plasma was higher than the activities observed in the other two species. Using this substrate, human and canine plasma enzyme activities were similar-unlike published data where hippuryl-histidyl-leucine was used as substrate.
Sigma-Aldrich
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly