  • Search Results
  • New formula herbal pellets demonstrate a uniform and stable release of the active ingredients in vitro.
New formula herbal pellets demonstrate a uniform and stable release of the active ingredients in vitro.
Acta poloniae pharmaceutica (2013-08-08)
Zivile Pranskuniene, Jurga Bernatoniene, Zenona Kalveniene, Ruta Masteikova, Tauras Mekas, Saule Velziene, Kostas Ivanauskas, Vytautas Suchockas, Inesa Mintauckiene, Arunas Savickas
ABSTRACT
The aim of this study was to evaluate the effect of different capsule filling manufacturing techniques and storage conditions on the release of the active ingredients from herbal capsules during the dissolution test in vitro. Different techniques for the preparation of the original mixture of dry extracts were applied, and subsequently capsules with six different fillings were prepared. The stability of the capsules was evaluated in different long-term storage conditions, registering changes in the water content (loss of drying), capsule disintegration time, and phenolic compounds dissolution test in vitro. The baseline of phenolic compounds release in the control capsules (filled with the mixture of the powder of dry herbal extracts) was the highest, compared to other capsule groups, yet during long-term storage, these capsules accumulated too much moisture, which impeded capsule disintegration time and phenolic compounds release. The study showed that moisture and temperature changes occurring during the storage of the preparation had a negative effect on the release of phenolic compounds from herbal capsules. Capsules filled with pellets demonstrated a uniform and stable release of the active ingredients in different long-term storage conditions, which indicates that the manufacturing technology of dry herbal extracts affects the stability of the active ingredients.
Sigma-Aldrich
Water, PCR Reagent
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, ACS reagent
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O