  • Search Results
  • Endohedral complexes of fullerene C60 with small convalent molecules (H2O, NH3, H2, 2H2, 3H2, 4H2, O2, O3) in the context of potential drug transporter system.
Endohedral complexes of fullerene C60 with small convalent molecules (H2O, NH3, H2, 2H2, 3H2, 4H2, O2, O3) in the context of potential drug transporter system.
Acta poloniae pharmaceutica (2013-08-08)
Marzena Medrek, Franciszek Pluciński, Aleksander P Mazurek
ABSTRACT
The fullerene C60 has chemical properties which seem to predestinate it to be effective transporter of drugs in biological system. To prove this, the DFT/B3LYP (6-31G*) calculations were performed especially in order to determine the structures and energies of the inclusion complexes of C60 with small molecules. It was found that the small molecule is more compact when it is located in the centre of the C60 cage than as isolated molecule. The calculated inclusion energies in case of: H2O, NH3, O2, O3, H2, 2H2, 3H2, 4H2 are: 1.84, 3.81, 3.75, 21.07, 1.97, 20.10, 47.78 and 77.54 kcal/mol, respectively. The charge transfer and the influence of the complexed small molecules on the electrostatic potential distribution inside and outside of the C60 cavity are discussed.
Sigma-Aldrich
Hydrogen, ≥99.99%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, PCR Reagent
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Fullerene-C60, 99.5%
Sigma-Aldrich
Fullerene-C60, 98%