  • Search Results
  • [A case of HER2-positive gastric cancer successfully treated with trastuzumab plus capecitabine plus cisplatin chemotherapy as third-line treatment].
[A case of HER2-positive gastric cancer successfully treated with trastuzumab plus capecitabine plus cisplatin chemotherapy as third-line treatment].
Gan to kagaku ryoho. Cancer & chemotherapy (2013-07-19)
Yoriko Murata, Tetsuya Matsuura, Kenji Kanoshima, Hiroki Kuwabara, Akiko Fuyuki, Wataru Tomeno, Reo Taniguchi, Takashi Uchiyama, Hitoshi Kuriyama, Yasuo Hata
ABSTRACT
A 67-year-old woman was diagnosed as having advanced gastric cancer(poorly differentiated adenocarcinoma)with multiple liver metastases. She had received combined S-1 plus cisplatin chemotherapy as first-line treatment and weekly paclitaxel chemotherapy as second-line treatment, however, both had eventually proved ineffective. Because the gastric cancer was HER2-positive, she was treated with trastuzumab plus capecitabine plus cisplatin(XP)chemotherapy as third-line treatment. The primary lesion and liver metastatic lesions were confirmed to show remarkable regression. The ToGA trial revealed increased efficacy of trastuzumab in first-line treatment of cancers showing high expression levels of the HER2- protein. This case suggested the increased efficacy of trastuzumab in third-line treatment. Neutropenia and hand foot syndrome of grade 2 were all reported adverse events. She could receive trastuzumab plus XP chemotherapy safely by dose reduction or dormancy temporarily of capecitabine.
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material