  • Search Results
  • Morphological evolution of silver nanoparticles and its effect on metal-induced chemical etching of silicon.
Morphological evolution of silver nanoparticles and its effect on metal-induced chemical etching of silicon.
Journal of nanoscience and nanotechnology (2013-07-19)
Seong-Ho Baek, Bo Hyun Kong, Hyung Koun Cho, Jae Hyun Kim
ABSTRACT
In this report, we have demonstrated the morphological evolution of the silver nanoparticles (AgNPs) by controlling the growth conditions and its effect on morphology of silicon (Si) during metal-induced electroless etching (MICE). Self-organized AgNPs with peculiarly shape were synthesized by an electroless plating method in a conventional aqueous hydrofluoric acid (HF) and silver nitrate (AgNO3) solution. AgNP nuclei were densely created on Si wafer surface, and they had a strong tendency to merge and form continuous metal films with increasing AgNO3 concentrations. Also, we have demonstrated that the fabrication of aligned Si nanowire (SiNW) arrays in large area of p-Si (111) substrates by MICE in a mixture of HF and hydrogen peroxide (H2O2) solution. We have found that the morphology of the initial AgNPs and oxidant concentration (H2O2) greatly influence on the shape of the SiNW etching profile. The morphological results showed that AgNP shapes were closely related to the etching direction of SiNWs, that is, the spherical AgNPs preferred to move vertical to the Si substrate, whereas non-spherical AgNPs changed their movement to the [100] directions. In addition, as the etching activity was increased at higher H2O2 concentrations, AgNPs had a tendency to move from the original [111] direction to the energetically preferred [100] direction.
Sigma-Aldrich
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silicon, powder, −60 mesh, 99.998% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Silicon, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.999% trace metals basis
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm