Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360)
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri™
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis