Urologic cancer risks for veterans exposed to Agent Orange.
Urologic nursing (2013-06-06)
Lori A Hoenemeyer
ABSTRACT
Agent Orange, an herbicide widely used during the Vietnam War, has been linked to various health risks, including urologic malignancy. Exposed veterans are at risk for prostate cancer and may be entitled to compensation if diagnosed with prostate cancer. Current research studies are aimed at mitigating prostate dysplasia and prostate cancer
Product Description
Supelco
2,4-D, certified reference material, TraceCERT®
Supelco
2,4-D, PESTANAL®, analytical standard
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
2,4,5-Trichlorophenoxyacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Supelco
2,4-D solution, certified reference material, 5000 μg/mL in acetonitrile
Supelco
2,4,5-T, PESTANAL®
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Supelco
2,3,7,8-Tetrachlorodibenzo-p-dioxin solution, analytical standard