  • Search Results
  • Effect of administration of vitamins C and E on fertilization capacity of rats exposed to noise stress.
Effect of administration of vitamins C and E on fertilization capacity of rats exposed to noise stress.
Noise & health (2013-05-22)
Ghasem Saki, Majid Jasemi, Ali Reza Sarkaki, Ali Fathollahi
ABSTRACT
The aims of this study were to evaluate the effects of administration of Vitamins C and E on fertilization capacity in rats exposed to noise stress. 40 adult male rats were randomly divided into 5 equal groups. Group 1 as controls who were not exposed to noise and groups 2-5 exposed to noise with 90-120 dB intensity and 300-350 Hz frequency from 7 pm to 7 am everyday for 50 days. Group 2 exposed to noise and did not receive Vitamins. Group 3 received vitamin C, Group 4 received Vitamin E. Group 5 received Vitamins C and E concomitantly. After 50 days, serum Follicle-stimulating hormone (FSH), Luteinizing hormone (LH) and testosterone were calculated. Then each rat was left with three female rats for mating. Pregnant females were sacrificed on the 19 th day of pregnancy and evaluated for the presence and number of viable, dead and absorbed fetuses. The level of FSH, LH and testosterone significantly decreased in rats exposed to noise (P < 0.05). By administration of Vitamins in groups 3-5 we observed that the level of hormones significantly increased in compared to group 2 (P < 0.05). The fertilization capacity of male rats in groups 3-5 significantly increased in compared to group 2 (P < 0.05). There was significant difference between groups 1 and 2 in case of fertilization capacity (P = 0.001). The data in this study strongly suggests a negative role for noise stress on level of FSH, LH and testosterone level and also fertilization capacity of male rats. To complement the information it is suggested that this research be done on human samples.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Follicle Stimulating Hormone from porcine pituitary
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Testosterone, ≥98%
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Follicle Stimulating Hormone from human pituitary, ~7,000 IU/mg (vs. W.H.O. reference preparation 83/575)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®