MilliporeSigma
Minireview: pH and synaptic transmission.
FEBS letters (2013-05-15)
Anne Sinning, Christian A Hübner
ABSTRACT
As a general rule a rise in pH increases neuronal activity, whereas it is dampened by a fall of pH. Neuronal activity per se also challenges pH homeostasis by the increase of metabolic acid equivalents. Moreover, the negative membrane potential of neurons promotes the intracellular accumulation of protons. Synaptic key players such as glutamate receptors or voltage-gated calcium channels show strong pH dependence and effects of pH gradients on synaptic processes are well known. However, the processes and mechanisms that allow controlling the pH in synaptic structures and how these mechanisms contribute to normal synaptic function are only beginning to be resolved.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder