  • Search Results
  • [Effect on different concentrations of exogenous hormones on baicalin in Scutellaria baicalensis callus].
[Effect on different concentrations of exogenous hormones on baicalin in Scutellaria baicalensis callus].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-01)
Gui-Xiang Wan, Lin Ma, Jian Zhang
ABSTRACT
To determine the content of baicalin in Scutellaria baicalensis callus induced by different doncentrations of exogenous hormones. HPLC system was adopted to determine baicalin in S. baicalensis callus. Chromatographic conditions: ODS column was adopted, with methanol-water-phosphate (47: 53: 0.2) as the mobile phase. The flow velocity was 1 mL x min(-1), the detective wavelength was 280 nm, and the temperature of column was room temperature. S. baicalensis callus induced by 6-BA 1.0 mg x L(-1) + NAA 0.5 mg x L(-1) showed the highest baicalin content, up to 49.78 mg x g(-1). The experiment is such a simple, rapid and stable method for determining the baicalin content that it can be used for determining the baicalin content in S. baicalensis callus.
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline
Sigma-Aldrich
Baicalin, 95%
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Supelco
2,4-D, certified reference material, TraceCERT®
Supelco
6-Benzylaminopurine, PESTANAL®, analytical standard
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
Sigma-Aldrich
Kinetin, BioReagent, suitable for plant cell culture, amorphous powder
Supelco
2,4-D, PESTANAL®, analytical standard
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Supelco
2,4-D solution, certified reference material, 5000 μg/mL in acetonitrile
Sigma-Aldrich
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)