  • Search Results
  • An injectable cell penetrable nano-polyplex hydrogel for localized siRNA delivery.
An injectable cell penetrable nano-polyplex hydrogel for localized siRNA delivery.
Biomaterials (2013-03-19)
Young-Min Kim, Mi-Ran Park, Soo-Chang Song
ABSTRACT
An approach for application of cell penetration to selective small interference RNA (siRNA) localized delivery system, cell penetrable nano-polyplex assembled hydrogel system, is presented. The cell penetrable nano-polyplex assembled hydrogelisprepared by protamine conjugation to poly(organophos-phazene) and inducement of nano-polyplexes with siRNAs. After an injection of cell penetrable nano-polyplex solution into the body, it turns into a gel due to thermosensitivity of poly(organo- phosphazene). The gel maintains up to 4 weeks and the released 30 nm-sized nano-polyplexes from the gel induces highly effective siRNA delivery due to cell penetration. Accordingly, the new system shows a high gene silencing efficiency on only the target site in long-term with a single injection.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer