Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Benzoic acid, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG