MilliporeSigma
A novel method for the preparation of a neurotoxic complex.
Analytical biochemistry (2013-01-22)
Bum Han Ryu, Eunjin Jang, Hansol Ju, T Doohun Kim
ABSTRACT
Neurodegenerative diseases such as Alzheimer's disease (AD) and Parkinson's disease (PD) can be attributed to the specific degeneration of neuronal cells in the brain. However, the natures and action modes of toxic species remain largely unknown. Here, we present a simple and fast method for the preparation of neurotoxic complex with α-synuclein, which is implicated in PD.
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Supelco
Oleic acid, Selectophore, ≥99.0%