  • Search Results
  • Evaluating pretreatment techniques for converting hazelnut husks to bioethanol.
Evaluating pretreatment techniques for converting hazelnut husks to bioethanol.
Bioresource technology (2012-12-19)
Yalçın Çöpür, Ayhan Tozluoglu, Melek Özkan
ABSTRACT
This study examined the suitability of husk waste for bioethanol production and compared pretreatment techniques with regard to their efficiencies. Results showed that 4% NaBH4 (90 min) delignified the highest amount of lignin (49.1%) from the structure. The highest xylan solubility (77.9%) was observed when samples were treated with 4% NaOH for 90 min. Pretreatment with NaOH and NaBH4, compared to H2O2 and H2SO4, resulted in selective delignification. The highest glucan to glucose conversion (74.4%) and the highest ethanol yield (52.6 g/kg husks) were observed for samples treated with 2% NaOH for 90 min.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether