MilliporeSigma
[Treatment and metaphylaxis of urate and mixed urolithiasis].
Urologiia (Moscow, Russia : 1999) (2012-10-19)
V P Avdoshin, M I Andriukhin, M N Israfilov, A V Annenkov, E A Atamanova
ABSTRACT
We estimated the efficacy of combined treatment and metaphylaxis of urate and mixed urolithiasis in 87 patients aged 18-87 years. The patients were divided into two groups: 68 patients of group 1 (the size of the concrements between 5-25 mm) have undergone parenteral litholysis of the drug trometamol N and metaphylaxis with biologically active additives prolit and urisan; 19 patients of group 2 have undergone extracorporeal shock-wave lithotripsy with parenteral litholysis and metaphylaxis by using biologically active additives prolit septo and urisan. Positive results were achieved in all the patients. In group 1 the concrements dissolved completely. In group 2 small fragments up to 4 mm eliminated after partial solution.
Product Description
Supelco
Trizma® base, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Pipemidic acid
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Tromethamine