MilliporeSigma
  • Structure of the O-polysaccharide of Azorhizobium caulinodans HAMBI 216; identification of 3-C-methyl-D-rhamnose as a component of bacterial polysaccharides.
Structure of the O-polysaccharide of Azorhizobium caulinodans HAMBI 216; identification of 3-C-methyl-D-rhamnose as a component of bacterial polysaccharides.
Carbohydrate research (2012-07-24)
Evelina L Zdorovenko, Olga A Valueva, Vadim V Kachala, Alexander S Shashkov, Yuriy A Knirel, Iwona Komaniecka, Adam Choma
ABSTRACT
The O-polysaccharide was obtained from the lipopolysaccharide of the stem-nodulating nitrogen-fixing bacterium Azorhizobium caulinodans HAMBI 216 and studied by sugar and methylation analyses along with (1)H and (13)C NMR spectroscopy. The polysaccharide was found to have a linear pentasaccharide repeating unit containing D-rhamnose and its rarely occurring 2-O-methyl (Rha2OMe) and 3-C-methyl (Rha3CMe) derivatives and having the following structure: →3)-α-D-Rhap2OMe-(1→2)-β-D-Rhap3CMe-(1→3)-α-D-Rhap-(1→2)-β-D-Rhap3CMe-(1→3)-α-D-Rhap-(1→ .
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG