  • Search Results
  • Dioxin/POPs legacy of pesticide production in Hamburg: part 1--securing of the production area.
Dioxin/POPs legacy of pesticide production in Hamburg: part 1--securing of the production area.
Environmental science and pollution research international (2012-07-11)
Roland Weber, Hans Gerhard Varbelow
ABSTRACT
α-Hexachlorocyclohexane (HCH), β-HCH, and γ-HCH (lindane) were recently included as new persistent organic pollutants (POPs) in the Stockholm Convention. Therefore, the chemicals need to be globally addressed, including the disposal of historic wastes. At most sites, the approximately 85% of HCH waste isomers were dumped. At a former lindane factory in Hamburg and some other factories the HCH, waste was recycled producing residues with high polychlorinated dibenzo-p-dioxin and dibenzofuran (PCDD/PCDF) levels. The soil and ground water under the former pesticide factory was/is highly contaminated with HCH (260 tons), chlorobenzenes (550 tons), and PCDD/PCDF (6 kg toxic equivalents (TEQ)). This contamination did not result from disposal operations but from spillages and leakages during the 30 years of the factory's production history. A containment wall has been constructed around the production area to prevent the dispersal of the pollutants. The ground water is managed by a pump and treat system. Over the last 15 years, approximately 10-30 tons of this pollution reservoir has been pumped and incinerated. For the contaminated production buildings, specific assessment and demolition technologies have been applied. In addition to their HCH waste isomer deposition, former lindane/HCH productions need to be assessed for possible recycling practice of HCH and related PCDD/PCDF contamination of the production area and buildings. Since such recycling activities have taken place at several factories in different countries, the experience of assessment and management of the described production area and contaminated buildings could be valuable. Such assessment could be addressed within the frame of the Stockholm Convention.
Product Description
Supelco
2,4,5-T, PESTANAL®