  • Search Results
  • Concise enantioselective construction of a bridged azatricyclic framework via domino semipinacol-Schmidt reaction.
Concise enantioselective construction of a bridged azatricyclic framework via domino semipinacol-Schmidt reaction.
Chemical communications (Cambridge, England) (2012-05-03)
Manohar Puppala, Annamalai Murali, Sundarababu Baskaran
ABSTRACT
A TiCl(4)-promoted domino semipinacol-Schmidt reaction of oxaspiropentane-azide provides an easy access to bridged azatricyclic ring systems, which possess the azaquaternary center, present in the immunosupressant FR901483 and platelet aggregation inhibitor daphlongeranine B.
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene