  • Search Results
  • A selective intramolecular transacylation of taxoids accompanying with the oxetane ring opening.
A selective intramolecular transacylation of taxoids accompanying with the oxetane ring opening.
Chemical & pharmaceutical bulletin (2012-03-03)
Feng Gao, Min Yu, Qiao-Hong Chen, Feng-Peng Wang
ABSTRACT
A selective intramolecular transacylation from C-4 to C-5 of taxoids, which occurred simultaneously with the oxetane D-ring opening and was promoted by TiCl4, was presented. The optimal condition was found to be treatment of substrates in double dose of dichloromethane (per mg of substrate dissolved in 2 mL DCM) with 1 eq of TiCl4 at 25°C for 10 min.
Sigma-Aldrich
Trimethylene oxide, 97%
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis