  • Search Results
  • Comparative analysis of gelatin scaffolds crosslinked by genipin and silane coupling agent.
Comparative analysis of gelatin scaffolds crosslinked by genipin and silane coupling agent.
International journal of biological macromolecules (2011-07-20)
C Tonda-Turo, P Gentile, S Saracino, V Chiono, V K Nandagiri, G Muzio, R A Canuto, G Ciardelli
ABSTRACT
Scaffolds based on gelatin (G) are considered promising for tissue engineering, able to mimic the natural extracellular matrix. G drawback is its poor structural consistency in wet conditions. Therefore, crosslinking is necessary to fabricate stable G scaffolds. In this work, a comparative study between the performance of two different crosslinkers, genipin (GP) and γ-glycidoxypropyltrimethoxysilane (GPTMS), is presented. Flat membranes by solvent casting and porous crosslinked scaffolds by freeze-drying were prepared. Infrared spectroscopy and thermal analysis were applied to confirm G chain crosslinking. Moreover, GP and GPTMS increased the stability of G in aqueous media and improved the mechanical properties. Crosslinking reduced the wettability, especially in the case of G_GPTMS samples, due to the introduction of hydrophobic siloxane chains. Both G_GP and G_GPTMS scaffolds supported MG-63 osteoblast-like cell adhesion and proliferation.
Sigma-Aldrich
(3-Glycidyloxypropyl)trimethoxysilane, ≥98%