  • Search Results
  • TiCl4-promoted cyclization reactions of aminoacetals and ethenetricarboxylates leading to nitrogen-containing heterocycles.
TiCl4-promoted cyclization reactions of aminoacetals and ethenetricarboxylates leading to nitrogen-containing heterocycles.
The Journal of organic chemistry (2011-06-21)
Shoko Yamazaki, Masachika Takebayashi
ABSTRACT
Lewis acid-catalyzed cyclization of aminoacetals 2 and triethyl ethenetricarboxylate (1a) has been examined. The reaction of 3-aminopropionaldehyde diethyl acetal (2a) and 1a in the presence of 1 equiv of TiCl(4) at room temperature gave 4-ethoxypiperidine-2,3,3-tricarboxylate 3a in 92% yield with a 2,4-diastereomer ratio of 1:1. The reaction in the presence of 3 equiv of TiCl(4) gave 2,4-trans-piperidine derivative 3a in 86% yield predominantly. The reaction of aminoacetaldehyde diethyl/dimethyl acetals 2c,d and 1a with 3 equiv of TiCl(4) gave 2,4-trans-4-pyrrolidine-2,3,3-tricarboxylates 5a,b predominantly.
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis