  • Search Results
  • Effect of shear force, solution pH and breakage period on characteristics of flocs formed by Titanium tetrachloride (TiCl4) and Polyaluminum chloride (PACl) with surface water treatment.
Effect of shear force, solution pH and breakage period on characteristics of flocs formed by Titanium tetrachloride (TiCl4) and Polyaluminum chloride (PACl) with surface water treatment.
Journal of hazardous materials (2011-02-12)
Y X Zhao, B Y Gao, H K Shon, J-H Kim, Q Y Yue
ABSTRACT
The growth, breakage and regrowth nature of flocs formed by Titanium tetrachloride (TiCl(4)) and polyaluminum chloride (PACl) was comparatively evaluated with surface water treatment. A series of jar experiments were conducted to investigate the impacts of different operating parameters such as shear force, solution pH and a breakage period on floc strength and re-aggregation potential. Results indicated that the responses of flocs to different operating parameters depend on the coagulant used. The ability of floc to resist breakage decreased with the increase of shear force and breakage period. Floc strength properties were also measured in response to increasing shear force, with the results suggesting that the order of floc strength was TiCl(4)>PACl. Floc regrowth of the two coagulants after exposure to high shear was limited, and flocs formed by TiCl(4) displayed weaker recoverability. The flocs generated in acid conditions were more recoverable than those generated in alkaline conditions no matter which coagulant was used.
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis