MilliporeSigma
Direct patterning of silanized-biomolecules on semiconductor surfaces.
Langmuir : the ACS journal of surfaces and colloids (2010-11-05)
Dorjderem Nyamjav, Richard C Holz
ABSTRACT
A novel approach to pattern silanized-biomolecules directly onto glass (SiO(x)) substrates via Dip-Pen nanolithography (DPN) and microcontact printing (μCP) is presented. Subsequent hybridization reactions of DPN patterned silanized-DNA with its complementary strands provide "proof-of-concept" that the patterned oligonucleotides maintain their biological activities. The fabrication strategy does not require premodification of substrates and offers a cheap and robust way to immobilize molecules on electronically important semiconductor surfaces.
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%