  • Search Results
  • An implicit solvent coarse-grained lipid model with correct stress profile.
An implicit solvent coarse-grained lipid model with correct stress profile.
The Journal of chemical physics (2010-06-03)
Alex J Sodt, Teresa Head-Gordon
ABSTRACT
We develop a coarse-grained parametrization strategy for lipid membranes that we illustrate for a dipalmitoylphosphatidylcholine bilayer. Our coarse-graining approach eliminates the high cost of explicit solvent but maintains more lipid interaction sites. We use a broad attractive tail-tail potential and extract realistic bonded potentials of mean force from all-atom simulations, resulting in a model with a sharp gel to fluid transition, a correct bending modulus, and overall very reasonable dynamics when compared with experiment. We also determine a quantitative stress profile and correct breakdown of contributions from lipid components when compared with detailed all-atom simulation benchmarks, which has been difficult to achieve for implicit membrane models. Such a coarse-grained lipid model will be necessary for efficiently simulating complex constructs of the membrane, such as protein assembly and lipid raft formation, within these nonaqueous chemical environments.
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%