  • Search Results
  • Harmonization of values for serum alkaline phosphatase catalytic activity concentration employing commutable calibration materials.
Harmonization of values for serum alkaline phosphatase catalytic activity concentration employing commutable calibration materials.
Clinica chimica acta; international journal of clinical chemistry (2010-03-17)
Giampaolo Cattozzo, Chiara Albeni, Carlo Franzini
ABSTRACT
Harmonization of results allows a more effective utilization of laboratory tests; we verified the feasibility of harmonizing serum alkaline phosphatase results by two methods. Patient sera (n=106) and candidate calibration materials (n=8) were analyzed by two methods, employing either diethanolamine (DEA) or 2-amino-2-methyl-1-propanol (AMP) as phosphate-accepting buffers. Results for patient sera by the DEA method were recalculated, with either a commutable or a non-commutable calibration material, both with values assigned by the AMP method. After calibration with the commutable material, the median intermethod difference (DEA-AMP) and ratio (DEA/AMP) dropped from 195 U/l to 0 U/l and from 2.47 to 1.00, respectively. When a non-commutable material was used the former became 124 U/l and the latter 1.94. After recalibration with the commutable material, linear regression and correlation analysis of DEA vs AMP values for the set of 106 patient sera gave: intercept=0.8 U/l; slope=0.997; and nonparametric correlation coefficient r=0.9995. Harmonization of alkaline phosphatase results by AMP and DEA methods is feasible when commutable calibration materials are used in the trueness transfer process.
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, ~5% Water, technical grade, 90%
Sigma-Aldrich
Diethanolamine hydrochloride, 98%
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, BioXtra, ≥95%
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, technical, ≥90% (GC)
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Supelco
Diethanolamine, analytical standard
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Supelco
Diethanolamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%