  • Search Results
  • The formation of straight and twisted filaments from short tau peptides.
The formation of straight and twisted filaments from short tau peptides.
The Journal of biological chemistry (2004-04-22)
Warren J Goux, Lauren Kopplin, Anh D Nguyen, Kathryn Leak, Marni Rutkofsky, Vasanthi D Shanmuganandam, Deepak Sharma, Hideyo Inouye, Daniel A Kirschner
ABSTRACT
We studied fibril formation in a family of peptides based on PHF6 (VQIVYK), a short peptide segment found in the microtubule binding region of tau protein. N-Acetylated peptides AcVYK-amide (AcVYK), AcIVYK-amide (AcPHF4), AcQIVYK-amide (AcPHF5), and AcV-QIVYK-amide (AcPHF6) rapidly formed straight filaments in the presence of 0.15 m NaCl, each composed of two laterally aligned protofilaments approximately 5 nm in width. X-ray fiber diffraction showed the omnipresent sharp 4.7-A reflection indicating that the scattering objects are likely elongated along the hydrogen-bonding direction in a cross-beta conformation, and Fourier transform IR suggested the peptide chains were in a parallel (AcVYK, AcPHF6) or antiparallel (AcPHF4, AcPHF5) beta-sheet configuration. The dipeptide N-acetyl-YK-amide (AcYK) formed globular structures approximately 200 nm to 1 microm in diameter. The polymerization rate, as measured by thioflavin S binding, increased with the length of the peptide going from AcYK --> AcPHF6, and peptides that aggregated most rapidly displayed CD spectra consistent with beta-sheet structure. There was a 3-fold decrease in rate when Val was substituted for Ile or Gln, nearly a 10-fold decrease when Ala was substituted for Tyr, and an increase in polymerization rate when Glu was substituted for Lys. Twisted filaments, composed of four laterally aligned protofilaments (9-19 nm width, approximately 90 nm half-periodicity), were formed by mixing AcPHF6 with AcVYK. Taken together these results suggest that the core of PHF6 is localized at VYK, and the interaction between small amphiphilic segments of tau may initiate nucleation and lead to filaments displaying paired helical filament morphology.
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)