MilliporeSigma
A novel method for screening beta-glucosidase inhibitors.
BMC microbiology (2013-03-19)
Sony Pandey, Ayinampudi Sree, Soumya S Dash, Dipti P Sethi
ABSTRACT
Few beta-glucosidase inhibitors have so far been reported from microorganisms due to the practical difficulties in performing the inhibition tests and subsequent interpretation of results. In an effort to investigate marine microbial extracts for β-glucosidase inhibitors, we developed a new protocol, using esculin as substrate in an agar plate based assay, to screen a large number of microbial extracts in a short span of time. With the new method, pale yellowish zones against the blackish brown background could be visually observed with more clarity in sample extracts where β-glucosidase inhibitor was present. The new method was compared with the closest existing method and established beyond doubt. This agar plate based procedure required about one hour for minimum 12 samples and the throughput increases with the size of the agar gel plate used. The new protocol was simple, rapid and effective in detecting beta-glucosidase inhibitors in microbial extracts.
Sigma-Aldrich
Bacteriological agar, for molecular biology
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized powder, ≥2 units/mg solid
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized powder, 10-30 units/mg solid
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type A, ash 5-6%
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized, powder, ≥4 U/mg
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type E, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Select agar
Millipore
Agar, for chromogenic media
Millipore
Agar, high purity, low ionic content, low gel strenght
Millipore
Agar, for membrane filtration, low gel strengh