  • Search Results
  • One hundred and twenty-seven cultured human tumor cell lines producing tumors in nude mice.
One hundred and twenty-seven cultured human tumor cell lines producing tumors in nude mice.
Journal of the National Cancer Institute (1977-07-01)
J Fogh, J M Fogh, T Orfeo
PMID327080
ABSTRACT
One hundred and twenty-seven cultured human tumor cell lines produced tumors after sc inoculation of 1-20 million cells into nude mice. They included 56 carcinoma lines, 14 sarcoma lines, and 57 lines from miscellaneous tumors and were all glucose-6-phosphate dehydrogenase type B. Twenty-nine percent of the lines produced tumors of 1 cm3 size within 1 month and 41% in the second month after inoculation. The histopathology correlated with the human tumor of origin in all cases.
Product Description
SW 626, 91091203
SW 403, 87071008
Sigma-Aldrich
CACO-2, Caucasian colon adenocarcinoma
CACO-2 Cell Line human, 86010202, from human colon(Caucasian colon adenocarcinoma)
SAOS-2 Cell Line human, primary osteogenic sarcoma, 89050205
CAKI 2, 93120819
SK-HEP-1, 91091816
CALU 1, 93120818