MilliporeSigma
Detection of acetylated alpha-tubulin by specific antibodies.
Methods in enzymology (1991-01-01)
M LeDizet, G Piperno
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture